Order Entry
Switzerland
ContactUsLinkComponent
Sodium hydroxide 0.111 M NaOH (0.111N) volumetric solution, for milk acidity analysis according to Dornic, Fluka™
Corrosive
Sodium hydroxide 0.111 M NaOH (0.111N) volumetric solution, for milk acidity analysis according to Dornic, Fluka™
Catalog # 34283-1L
CAS Number:  
Corrosive
Sodium hydroxide 0.111 M NaOH (0.111N) volumetric solution, for milk acidity analysis according to Dornic, Fluka™
Catalog # 34283-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Vapor Pressure:
    3 mmHg ( 37 °C)
  • Factor:
    0.999 - 1.001

Specification Test Results

About this item

 :
 :40 g/mol
Boiling Pt:100 °C (1.013 hPa)
Density:1.000 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-791-2
UN:1824
ADR:8,III
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548