Order Entry
Switzerland
ContactUsLinkComponent
Sodium hydroxide 0.05 mol/l (0.05 N) in aqueous solution, AVS TITRINORM volumetric solution
Sodium hydroxide 0.05 mol/l (0.05 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 31630.303
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium hydroxide 0.05 mol/l (0.05 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 31630.303
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2 L
  • Seq:
    5086

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    0.0499 - 0.0501 mol/l

Specification Test Results

About this item

 :
 :40 g/mol
Density:1 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,III
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548