ContactUsLinkComponent
Potassium hydroxide 0.1 mol/l (0.1 N) in ethanol, Titripur® Reag. Ph. Eur., Reag. USP volumetric solution, Supelco®
Potassium hydroxide 0.1 mol/l (0.1 N) in ethanol, Titripur® Reag. Ph. Eur., Reag. USP volumetric solution, Supelco®
Supplier:  Merck
Certificates
Potassium hydroxide 0.1 mol/l (0.1 N) in ethanol, Titripur® Reag. Ph. Eur., Reag. USP volumetric solution, Supelco®
Supplier:  Merck
Certificates
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Form:
    liquid
  • Amount-of-substance concentration:
    0.0995-0.1005
  • Measurement uncertainty:
    +/- 0.0003
  • Traceability:
    NIST SRM