Hydrobromic acid 47%, GPR RECTAPUR®
Supplier: VWR Chemicals
About this item
Synonyms: Hydrogen bromide
- CAS Number:10035-10-6
- Formula:HBr
- MDL Number:MFCD00011323
- Molecular Weight:80.91 g/mol
- Density:1.47 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:233-113-0
- UN:1788
- ADR:8,II
2 Options Available Below
Product Details & Documents
Synonyms: Hydrogen bromide
- CAS Number:10035-10-6
- Formula:HBr
- MDL Number:MFCD00011323
- Molecular Weight:80.91 g/mol
- Density:1.47 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:233-113-0
- UN:1788
- ADR:8,II
Specifications for Product Family
Specification Test Results
- Assay:Min. 47 %
- Phosphate, Phosphite (as PO4):max 0.002%
- Sulphate and sulphite (as SO4):max 0.005%
- Heavy metals (as Pb):max 0.001%
- Fe (Iron):max 0.0005%



