Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Acrylamide 40% in aqueous solution

Acrylamide 40% in aqueous solution

Toxic
Health
Marketsource
Supplier:   GERBU BIOCHEMICALS
Acrylamide 40% in aqueous solution
Supplier:  GERBU BIOCHEMICALS
Marketsource
Certificates
Acrylamide 40% in aqueous solution
Acrylamide 40% in aqueous solution

3 Options Available Below

Product Details & Documents
Synonyms:  Prop-2-enamide
  • CAS Number:79-06-1
  • Formula:C₃H₅NO
  • MDL Number:MFCD00008032
  • Molecular Weight:71.08 g/mol
  • EINECS:201-173-7
  • UN:3426
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L

    Specifications:
    Marketsource
    Marketsource
  • 6 L
    Log in for availability
    Log in for availability
    1 * 6 L
    1 * 6 L

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.