Acrylamide 30% in aqueous solution
Supplier: GERBU BIOCHEMICALS
Certificates
About this item
Synonyms: Prop-2-enamide
CAS Number:79-06-1
:
Formula:C₃H₅NO
MDL Number:MFCD00008032
:71.08 g/mol
EINECS:201-173-7
UN:3426
ADR:6.1,III
:GHS06
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together