Order Entry
Switzerland
ContactUsLinkComponent
Hydrochloric acid 6 mol/l (6 N), BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Hydrochloric acid 6 mol/l (6 N), BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 7710.9010
Supplier:  Avantor
CAS Number:  
Hydrochloric acid 6 mol/l (6 N), BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 7710.9010
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    10 L
  • Seq:
    5214

Specifications

Specification Test Results

  • Appearance of solution:
    clear, colourless
  • Normality (N):
    5.9 - 6.1

Specification Test Results

About this item

 :
Density:1.06 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-595-7
ADR:8,II
 :36.46 g/mol
UN:1789
 :GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324