Order Entry
Switzerland
ContactUsLinkComponent
Hydrochloric acid 6 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Irritant
Corrosive
Hydrochloric acid 6 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Catalog # 5619-02
Supplier:  Avantor
CAS Number:  
undefined
Irritant
Corrosive
Hydrochloric acid 6 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Catalog # 5619-02
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    PE bottle, narrow mouth
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    5.95 - 6.05
  • Normality (eq/L):
    5.95 - 6.05
  • Sulfate (SO₄):
    ≤ 1 ppm
  • Ammonium (NH₄):
    ≤ 3 ppm
  • Trace Impurities - Heavy Metals:
    ≤ 0.5 ppm
  • Trace Impurities - Iron (Fe):
    ≤ 0.1 ppm
  • Appearance:
    Passes Test

Specification Test Results

About this item

 :
Density:1.06 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
ADR:8,II
 :Management of Change (MOC) category = N
 :36.46 g/mol
 :GHS07
 :GHS05
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324