Order Entry
Switzerland
ContactUsLinkComponent
N-Lauroylsarcosine sodium salt 30% in aqueous solution
Toxic
N-Lauroylsarcosine sodium salt 30% in aqueous solution
Catalog # A3457.0500
Supplier:  PanReac AppliChem
CAS Number:  
undefined
Toxic
N-Lauroylsarcosine sodium salt 30% in aqueous solution
Catalog # A3457.0500
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :293.38 g/mol
EINECS:205-281-5
 :GHS06
Storage Temperature:Ambient
CAS Number:137-16-6
Formula:H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa
MDL Number:MFCD00042728