
Hydrochloric acid 0.714 mol/l (N/1.4)
Supplier: PROLABO CMP
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
ADR:8,II
:36.46 g/mol
EINECS:231-595-7
UN:1789
:GHS05
Storage Temperature:Ambient
Frequently Bought Together