Order Entry
Switzerland
ContactUsLinkComponent
EDTA disodium salt 0.01 mol/l (0.02 N) in aqueous solution, AVS TITRINORM volumetric solution
EDTA disodium salt 0.01 mol/l (0.02 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 160226G
Supplier:  VWR Chemicals
CAS Number:  
undefined
EDTA disodium salt 0.01 mol/l (0.02 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 160226G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic container
  • Pk:
    5 L
  • Seq:
    5190

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    0.00998 - 0.01002 mol/l
  • NIST traceable:
    Confirmed

Specification Test Results

About this item

 :
 :336.21 g/mol
Storage Temperature:Ambient
EINECS:205-358-3
CAS Number:139-33-3
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672