
Oxalic acid 0.5 M (1 N) volumetric solution, Fluka™
Catalog # 35295-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Factor:0.999 - 1.001
Specification Test Results
About this item
:
:90.04 g/mol
Boiling Pt:100 °C
Density:1.02 g/cm3
Storage Temperature:Ambient
EINECS:205-634-3
ADR:8,III
:GHS05
CAS Number:144-62-7
Formula:C₂H₂O₄
MDL Number:MFCD00002573