Order Entry
Switzerland
ContactUsLinkComponent
Oxalic acid 0.05 mol/l (0.1 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Oxalic acid 0.05 mol/l (0.1 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 7141.1000
Supplier:  Avantor
CAS Number:  
Oxalic acid 0.05 mol/l (0.1 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 7141.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Titer (mol/l):
    0.0497 - 0.0503

Specification Test Results

About this item

 :
 :90.04 g/mol
Storage Temperature:Ambient
EINECS:205-634-3
ADR:8,III
CAS Number:144-62-7
Formula:C₂H₂O₄
MDL Number:MFCD00002573