Oxalic acid 0.05 mol/l (0.1 N) in aqueous solution volumetric solution
Catalog # 35622.K2
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:90.04 g/mol
Storage Temperature:Ambient
EINECS:205-634-3
ADR:8,III
CAS Number:144-62-7
Formula:C₂H₂O₄
MDL Number:MFCD00002573