Order Entry
Switzerland
ContactUsLinkComponent
Oxalic acid standard solution 0.1 N
Oxalic acid standard solution 0.1 N
Certificates
undefined
Oxalic acid standard solution 0.1 N

Specification Test Results

  • Appearance:
    Clear colorless Solution
  • Strength:
    0.0998 to 0.1002 N (20 °C)
  • Melting point:
    116 to 119 °C
  • GC:
    ≥96.0 %
  • NMR (1H-NMR):
    Consistent with structure

Specification Test Results

Related Information

Frequently Bought Together