Glutaraldehyde 25% in aqueous solution for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Glutaral, Glutardialdehyde, 1,5-Pentanedial
CAS Number:111-30-8
:
Formula:C₅H₈O₂
MDL Number:MFCD00007025
:100.12 g/mol
Boiling Pt:101 °C (1013 hPa)
Melting Pt:–6 °C
Density:1.06 g/cm³ (20 °C)
Flash Pt:>71 °C
UN:2922
ADR:8 (6.1),II
:M-Clarity™ quality level = MQ200
EINECS:203-856-5
:GHS05
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-6 °C
- Boiling Point:100.5 °C (1013 hPa)
- Vapor Pressure:0.27 hPa (20 °C)
- Density:1.06 g/cm3 (20 °C)
- pH:3.0 (H2O, 20 °C)
- Ignition Point:275 °C
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
Related Information
Frequently Bought Together