Order Entry
Switzerland
ContactUsLinkComponent
Glutaraldehyde 25% in aqueous solution, Sigma-Aldrich®
Glutaraldehyde 25% in aqueous solution, Sigma-Aldrich®
Catalog # 354400-500
CAS Number:  
Glutaraldehyde 25% in aqueous solution, Sigma-Aldrich®
Catalog # 354400-500
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Glutaral, Glutardialdehyde, 1,5-Pentanedial
 :
 :100.12 g/mol
Boiling Pt:101 °C (1013 hPa)
Melting Pt:–6 °C
Density:1.06 g/cm³ (20 °C)
Flash Pt:>71 °C
Storage Temperature:Refrigerator
EINECS:203-856-5
UN:2927
ADR:6.1,II
 :GHS06
 :GHS05
 :GHS08
 :GHS09
CAS Number:111-30-8
Formula:C₅H₈O₂
MDL Number:MFCD00007025