Order Entry
Switzerland
ContactUsLinkComponent
 
Acetic acid 1 mol/l (1 N), AVS TITRINORM volumetric solution
  30010.292
 :  
undefined
Acetic acid 1 mol/l (1 N), AVS TITRINORM volumetric solution
  30010.292
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Titer (20°C; real value 0.2 % accuracy):
    0.998 - 1.002 mol/l
  • NIST traceable:
    Confirmed

 

 

 :
 :60.05 g/mol
 :Ambient
 :200-580-7
 :64-19-7
 :H₃CCOOH
 :MFCD00036152