
(±)-1-Octen-3-ol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3391-86-4
:
Formula:C₈H₁₆O
MDL Number:MFCD00004589
Boiling Pt:174 °C (760 mmHg)
Melting Pt:-49 °C
Density:0.837 g/cm³
Flash Pt:68 °C
Storage Temperature:Ambient
EINECS:222-226-0
ADR:6.1,III
:128.21 g/mol
:GHS07
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4340 to 1.4420 (20°C, 589 nm)
- Density:(20°C) 0.832 to 0.842 kg/l
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together