
(±)-α-Methoxyphenylacetic acid 99%
:7021-09-2
:
:C₉H₁₀O₃
:MFCD00064217
:166.18 g/mol
:165 °C (24 hPa)
:68…72 °C
:Ambient
:230-300-9
:GHS07
- Appearance:Off-white to light beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:68 to 72 °C
- Titration with NaOH:≥98.5 %
Frequently Bought Together