Order Entry
Switzerland
ContactUsLinkComponent
 
Health
Environment
(±)-3,5,5-Trimethyl-1-hexanol, TECHNICAL
Catalog # 262134R
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Environment
(±)-3,5,5-Trimethyl-1-hexanol, TECHNICAL
Catalog # 262134R
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • IR Spectrum:
    Passes test
  • Density (20/4):
    0.826 - 0.831

Specification Test Results

About this item

 :
Boiling Pt:193…194 °C (1013 hPa)
Melting Pt:–70 °C
Density:0.83 g/cm³ (20 °C)
Flash Pt:80 °C (closed cup)
Storage Temperature:Ambient
EINECS:222-376-7
UN:3082
ADR:9,III
 :144.26 g/mol
 :GHS08
 :GHS09
CAS Number:3452-97-9
Formula:(CH₃)₃CCH₂CH(CH₃)CH₂CH₂OH
MDL Number:MFCD00036138