Order Entry
Switzerland
ContactUsLinkComponent
(±)-1-Methoxy-2-propanol ≥98%, GPR RECTAPUR®
Flammable
Irritant
(±)-1-Methoxy-2-propanol ≥98%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Irritant
(±)-1-Methoxy-2-propanol ≥98%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 98 %
  • Acidity:
    Max. 0.0003 meq/g
  • Water:
    Max. 0.1 %

Specification Test Results

Frequently Bought Together