Order Entry
Switzerland
ContactUsLinkComponent
Diethyl ether stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Diethyl ether stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Diethyl ether stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay (by GC):
    min. 99.5 %
  • Alcohol (C2H5OH):
    max. 0.01 %
  • Carbonyl Compounds (as HCHO):
    max. 0.001 %
  • Color (APHA):
    max. 10
  • Density (g/ml) at 20°C:
    0.712 - 0.714
  • Foreign Odor:
    passes test
  • Peroxide (as H2O2):
    max. 3 ppm
  • Residue after Evaporation:
    max. 0.001 %
  • Substances Darkened by H2SO4:
    passes test
  • Titrable Acid (meq/g):
    max. 0.0002
  • Water (H2O):
    max. 0.01 %
  • Aluminium (Al):
    max. 0.5
  • Boron (B):
    max. 0.02
  • Barium (Ba):
    max. 0.1
  • Calcium (Ca):
    max. 0.5
  • Cadmium (Cd):
    max. 0.05
  • Cobalt (Co):
    max. 0.02
  • Chromium (Cr):
    max. 0.02
  • Copper (Cu):
    max. 0.02
  • Iron (Fe):
    max. 0.1
  • Magnesium (Mg):
    max. 0.1
  • Manganese (Mn):
    max. 0.02
  • Nickel (Ni):
    max. 0.02
  • Lead (Pb):
    max. 0.1
  • Tin (Sn):
    max. 0.1
  • Zinc (Zn):
    max. 0.1

Specification Test Results

Related Information

Frequently Bought Together