Order Entry
Switzerland
ContactUsLinkComponent
 
Diethyl ether, anhydrous 99+% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
 
undefined
Diethyl ether, anhydrous 99+% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.

 

  • Appearance:
    Clear liquid
  • Water:
    ≤0.03 % (Coulometric)
  • Boiling point:
    34 to 35 °C
  • Density:
    (20/20 °C) 0.713 to 0.715
  • GC:
    ≥99.0 %
  • Color scale:
    ≤10 APHA
  • Stabilizer:
    ≤10 ppm BHT
  • Residue after evaporation:
    ≤0.001 %
  • Carbonyl (as HCHO):
    ≤0.001 %
  • Titrable acid:
    ≤0.0002 meq/g
  • Ethanol:
    (limit about 0.05%) Passes test
  • Peroxides:
    ≤1 ppm (as H2O2)

 

 

Frequently Bought Together