DL-Methionine
Catalog # MOLE13855549-1KG
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:149.21 g/mol
Merck Index:13,06004
Melting Pt:∼136 °C
Density:1.34 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-432-1
CAS Number:59-51-8
Formula:C₅H₁₁NO₂S
MDL Number:MFCD00063096