

Chloroform, extra dry over Molecular Sieve 99.9% stabilised, AcroSeal®
364321000.
:
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 %
- GC:≥99.85 %
- Stabilizer:50 to 150 ppm Amylene
:
:119.38 g/mol
:13,02160
:61 °C (760 mmHg)
:–63 °C
:1.48 g/cm³
:Ambient
:200-663-8
:1888
:6.1,III
:05-2114614906-45
:GHS06
:GHS08
:67-66-3
:CHCl₃
:MFCD00000826