

Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-09-2
:
Formula:CH₂Cl₂
MDL Number:MFCD00000881
:84.93 g/mol
Merck Index:12,06140
Boiling Pt:39.8 °C (1013 hPa)
Melting Pt:–97 °C
Density:1.325 g/cm³
UN:1593
ADR:6.1,III
:Stabilised with amylene
EINECS:200-838-9
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.02 %
- GC:≥99.8 %
- UV:at 233 nm A: ≤1
- Color scale:≤10 APHA
- Stabilizer:About 100 ppm Amylene
- Residue after evaporation:≤0.0002 %
- Acidity:≤0.0001 meq/g
- Filtered to 0.2 micrometer:Filtered
- Free chlorine:≤0.0005 %
Specification Test Results
Frequently Bought Together