Order Entry
Switzerland
ContactUsLinkComponent
Dow Corning® FS 1265/10,000 cS chemically inert fluid
Dow Corning® FS 1265/10,000 cS chemically inert fluid
Catalog # 630267H
Supplier:  DOW CORNING
CAS Number:  
Dow Corning® FS 1265/10,000 cS chemically inert fluid
Catalog # 630267H
Supplier:  DOW CORNING
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 kg
  • Seq:
    4114

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:63148-56-1