Tween® 20 (Polysorbate)
Marketsource
Supplier: NATIONAL DIAGNOSTICS
Jump to:
About this item
Synonyms: Polyoxyethylene sorbitan monolaurate
- CAS Number:9005-64-5
- Formula:C₅₈H₁₁₄O₂₆
- MDL Number:MFCD00165986
- Molecular Weight:1227.53 g/mol
- Density:1.1 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:500-018-3
2 Options Available Below
Product Details & Documents
Synonyms: Polyoxyethylene sorbitan monolaurate
- CAS Number:9005-64-5
- Formula:C₅₈H₁₁₄O₂₆
- MDL Number:MFCD00165986
- Molecular Weight:1227.53 g/mol
- Density:1.1 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:500-018-3
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



