Cold pack, HypaCool
HypaCool instant cold pack, standard, 230×140 mm
SAFIQ2290
This instant single use cold pack is ideal for use in the event of an emergency or injury, delivering fast first aid to those who need it. It also helps to control bleeding, inflammation, swelling and pain.
- Quick, easy and mess-free way of providing cold treatment during the acute stages of injury
- Quality cold pack activates instantly when squeezed
- No need to place in the freezer
- Instant, soothing cold therapy
- Recommended addition to any first aid kit
Weight: 200 g
- Description:HypaCool instant cold pack, standard, 230×140 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



