Order Entry
Switzerland
ContactUsLinkComponent
Polyethylene glycol 400 for synthesis, Sigma-Aldrich®
Polyethylene glycol 400 for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Polyethylene glycol 400 for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    4 - 8 °C
  • Molar Mass:
    8000 g/mol
  • Boiling Point:
    200 °C (1013 hPa)
  • Vapor Pressure:
     - 0.1 hPa (20 °C)
  • Flash Point:
    240 °C
  • Density:
    1.13 g/cm3 (20 °C)
  • pH:
    5 - 7 (100 g/l, H2O, 20 °C)
  • Ignition Point:
    360 °C DIN 51794
  • Viscosity kinematic:
    97 - 110 mm2/s (20 °C)

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together