Order Entry
Switzerland
ContactUsLinkComponent
Polyethylene glycol 10,000 for synthesis, Sigma-Aldrich®
Polyethylene glycol 10,000 for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Polyethylene glycol 10,000 for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    500 g/l (20 °C)
  • Melting Point:
    58 - 63 °C
  • Molar Mass:
    9000 g/mol
  • Bulk Density:
    400 - 500 kg/m3
  • Vapor Pressure:
     - 0.01 hPa (20 °C)
  • Flash Point:
    265 °C
  • Density:
    1.2 g/cm3 (20 °C)
  • pH:
    5 - 7 (100 g/l, H2O, 20 °C)
  • Ignition Point:
    340 °C
  • Viscosity kinematic:
    500 - 900 mm2/s (20 °C)

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together