Order Entry
Switzerland
ContactUsLinkComponent
VWR® ARISTAR®, Silver Standard Solution, 10000 mg/L Ag in dil. Nitric Acid (from AgNO₃), Standard for ICP
VWR® ARISTAR®, Silver Standard Solution, 10000 mg/L Ag in dil. Nitric Acid (from AgNO₃), Standard for ICP
Catalog # 456032M
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, Silver Standard Solution, 10000 mg/L Ag in dil. Nitric Acid (from AgNO₃), Standard for ICP
Catalog # 456032M
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Silver
  • Symbol:
    Ag

Specifications

About this item

AgNO₃ in HNO₃ 2 to 5%.

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Supplied with certificate of analysis.