Silver standard solution, 1000 mg/L Ag in dil. nitric acid (from AgNO₃) Certipur® standard for ICP, Supelco®
1.70352.0100
: Merck
:
- Concentration:1000 mg/L
- Matrix:Nitric acid dil.
- Pack type:Plastic bottle
- Pk:100 mL
- Environmentally Preferable:
- M-Clarity quality level:MQ100
- Element:Silver
- Symbol:Ag
: Traceable to SRM from NIST.