VWR® ARISTAR®, Silver Standard Solution, 1000 mg/L Ag in dil. Nitric Acid (from AgNO₃), Standard for ICP
Supplier: VWR Chemicals
Certificates
About this item
AgNO₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.
Limited Time Sale
Select items below are currently on sale
Specifications
- Environmentally Preferable:
Specifications
Related Information
Certificates
Search by Lot Number125 ml Plastic bottle for liquids DIN 24 (ZPACK15150125) 500 ml Plastic bottle for liquids DIN 28 (ZPACK15150500)
Packaging Information
VWR® for Calibrations VWR® Trace Analysis.pdf
Product Literature
Frequently Bought Together