Order Entry
Switzerland
ContactUsLinkComponent
VWR® ARISTAR®, Magnesium Standard Solution, 1000 mg/L Mg in dil. Nitric Acid (from Mg), Standard for Ion Chromatography
VWR® ARISTAR®, Magnesium Standard Solution, 1000 mg/L Mg in dil. Nitric Acid (from Mg), Standard for Ion Chromatography
Catalog # 458212W
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, Magnesium Standard Solution, 1000 mg/L Mg in dil. Nitric Acid (from Mg), Standard for Ion Chromatography
Catalog # 458212W
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Magnesium
  • Symbol:
    Mg

Specifications

About this item

Mg in H₂O tr.HNO₃.

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Certifications: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Delivery: Supplied with certificate of analysis.