Chromium standard solution, 10000 mg/L Cr in dil. nitric acid (from Cr(NO₃)₃) Certipur® standard for ICP, Supelco®
Katalognr. 1.70374.0100
Supplier: Merck
CAS Number:
Specifications
- Concentration:10000 mg/L
- Matrix:2-3% HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- M-Clarity quality level:MQ100
- Element:Chromium
- Symbol:Cr
Specifications
About this item
Certifications: Traceable to SRM from NIST.