Order Entry
Switzerland
ContactUsLinkComponent
 
Arsenic standard solution, concentrate, 1000 mg As in water (from As₂O₅) Titrisol® standard for AAS, Supelco®
  1.09939.0001
 :  Merck
 :  
Arsenic standard solution, concentrate, 1000 mg As in water (from As₂O₅) Titrisol® standard for AAS, Supelco®
  1.09939.0001
 :  Merck
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • M-Clarity quality level:
    MQ100
  • Element:
    Arsenic
  • Symbol:
    As

 

 

 : Traceable to SRM from NIST.