Order Entry
Switzerland
ContactUsLinkComponent
 
Human albumin (from serum)
  CHMCAB3391
 :  Chemicon
 :  
Human albumin (from serum)
  CHMCAB3391
 :  Chemicon
 :  

 

  • Pk:
    100 µG
  • CAS No.:
    70024-90-7
  • Source:
    Serum
  • Species:
    Human
  • Storage conditions:
    Refrigerator
  • Protein synonyms:
    Human Serum Albumin|HSA|serum albumin|HAS|serum of albumin
  • Protein/peptide name:
    Albumin