Auftragserfassung
Switzerland
ContactUsLinkComponent
Kanadabalsam, TECHNICAL
Kanadabalsam, TECHNICAL
Catalog # 21776.183
Supplier:  VWR Chemicals
CAS Number:  
undefined
Kanadabalsam, TECHNICAL
Catalog # 21776.183
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Packung:
    Glasflasche
  • VE:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Identification:
    Passes test

Specification Test Results

About this item

 :
Merck Index:13,00948
Density:0.915 g/cm³ (20 °C)
Flash Pt:62 °C
Storage Temperature:Raumtemperatur
EINECS:232-362-2
UN:1993
ADR:3,III
CAS Number:8007-47-4
MDL Number:MFCD00132800