Starch (from wheat), powder, TECHNICAL
Catalog # 21146.290
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- IR Spectrum:Passes test
Specification Test Results
About this item
:
Density:1.5 g/cm³ (20 °C)
:162.14 g/mol
Storage Temperature:Ambient
EINECS:232-679-6
CAS Number:9005-25-8
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00082026