Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Agarose

Agarose

Marketsource
Supplier:   Roth Carl
Agarose
Supplier:  Roth Carl
Marketsource
Certificates
Agarose

4 Options Available Below

Product Details & Documents
  • CAS Number:9012-36-6
  • MDL Number:MFCD00081294
  • Melting Pt:60…90 °C
  • Storage Temperature:Ambient
  • EINECS:232-731-8
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - SFr. 31.80
    SFr. 31.80
    1 * 10 g
       SFr. 31.80

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - SFr. 174.00
    SFr. 174.00
    1 * 100 g
       SFr. 174.00

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - SFr. 714.00
    SFr. 714.00
    1 * 500 g
       SFr. 714.00

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - SFr. 1’194.00
    SFr. 1’194.00
    1 * 1 kg
       SFr. 1’194.00

    Specifications:
    Marketsource
    Marketsource