Agarose MS8 molecular sieve grade, for small DNA fragments
:9012-36-6
:
:MFCD00081294
:60…90 °C
:Ambient
:232-731-8
Frequently Bought Together
Methanol ≥98.5%, TECHNICAL
: VWR Chemicals
Ethyl acetate ≥99%, GPR RECTAPUR®
: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Ethanol 96% vol Ph. Eur.
: VWR Chemicals