Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Agar

Supplier:   EUROCLONE
Agar
Supplier:  EUROCLONE
Certificates
Agar

3 Options Available Below

Product Details & Documents
Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • MDL Number:MFCD00081288
  • Merck Index:13,00184
  • Melting Pt:90 °C
  • Storage Temperature:Ambient
  • EINECS:232-658-1
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • 1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT

  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.