Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Agar
Agar
Supplier:  PanReac AppliChem
Certificates
Agar

2 Options Available Below

Product Details & Documents
Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • MDL Number:MFCD00081288
  • Merck Index:13,00184
  • Melting Pt:90 °C
  • Storage Temperature:Ambient
  • EINECS:232-658-1

Documents

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - SFr. 213.00
    SFr. 213.00
    1 * 1 kg
       SFr. 213.00

  • 5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - SFr. 850.00
    SFr. 850.00
    1 * 5 kg
       SFr. 850.00