Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Agar, powder

Agar, powder

Agar, powder
Certificats
Agar, powder

3 variantes

Product Details & Documents

CAS No.: 9002-18-0 Molecular Formula: (C12H18O9)X Physical Form: Powder Appearance: White to cream Odour: Odourless Merck Reference: 14,184 MDL No.: MFCD00081288

Synonymes:  Agar Agar
  • Numéro CAS:9002-18-0
  • Numéro MDL:MFCD00081288
  • Index Merck:14,184
  • Point de fusion:90 °C
  • Température de stockage:Ambient
  • EINECS:232-658-1
Product Family Options

Informations sur le produit

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.