Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Cellulose, Supelco®

Cellulose, Supelco®

Glass bottle for solids, 1 kg
Catalog # 22182-1KG
Supplier:   Merck
Cellulose, Supelco®
Catalog # 22182-1KG
Supplier:  Merck
CAS Number:  9004-34-6
Cellulose, Supelco®
Cellulose, Supelco®

Glass bottle for solids, 1 kg

CAS Number:  9004-34-6
Price
SFr. 313.00
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Microcrystallinecellulose
  • CAS Number:9004-34-6
  • Formula:(C₁₂H₂₂O₁₁)n
  • MDL Number:MFCD00081512
  • Molecular Weight:342.3 g/mol
  • Merck Index:13,01977
  • Melting Pt:260…270 °C
  • Density:1.27…1.6 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ100
  • Storage Temperature:Ambient
  • EINECS:232-674-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications