Order Entry
Switzerland
ContactUsLinkComponent
Cellulose MN 2100 adsorbents for column chromatography
Cellulose MN 2100 adsorbents for column chromatography
Supplier:  MACHEREY-NAGEL
Certificates
undefined
Cellulose MN 2100 adsorbents for column chromatography
Supplier:  MACHEREY-NAGEL
Certificates

Specification Test Results

  • Residue on Ignition:
    <10000 ppm
  • Fe:
    <2 ppm
  • Cu:
    <1 ppm
  • P:
    <2 ppm
  • Dichloromethane Extract:
    <0.15%

Specification Test Results

Related Information

Frequently Bought Together