Order Entry
Switzerland
Orders LinkContactUsLinkComponent
D-(-)-Fructose Ph. Eur.

D-(-)-Fructose Ph. Eur.

D-(-)-Fructose Ph. Eur.
Supplier:  VWR Chemicals
Certificates
D-(-)-Fructose Ph. Eur.

3 Options Available Below

Product Details & Documents
Synonyms:  D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose
  • CAS Number:57-48-7
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00148910
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,04295
  • Boiling Pt:552 °C (1013 hPa)
  • Melting Pt:100…110 °C
  • Density:1.59 g/cm³ (20 °C. OECD 109)
  • Storage Temperature:Ambient
  • EINECS:200-333-3
Specifications for Product Family

Specification Test Results

  • Appearance:
    Colourless crystals
  • Identification A:
    Passes test
  • Identification B:
    Passes test
  • Identification C:
    Passes test
  • Identification D:
    Passes test
  • Appearance of solution:
    Passes test
  • Acidity or alkalinity:
    Passes test
  • Spec. opt. rotation (calc. on anhydrous):
    -93.5 to -91 deg
  • Foreign sugars:
    Passes test
  • 5-Hydroxymethylfurfural + related subst.:
    Passes test
  • Ba (Barium):
    Passes test
  • Lead in sugars:
    Max. 0.5 ppm
  • Water:
    Max. 0.5 %
  • Sulphated ash:
    Max. 0.1 %
  • Residual solvents:
    Passes test
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.