D-(+)-Xylose
Glass bottle for solids, 100 g
Catalog # ROTH5537.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
- CAS Number:58-86-6
- Formula:C₅H₁₀O₅
- MDL Number:MFCD00151475
- Molecular Weight:150.13 g/mol
- Merck Index:13,10142
- Boiling Pt:331 °C (1013 hPa)
- Melting Pt:154 °C
- Density:1.525 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-400-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Cat. No.:ROTH5537.1



